Erie SoftCIDE® Hand Wash
Supplier: Erie Scientific
21128-04-001
490007-306CS 385.6 USD
490007-306
Erie SoftCIDE® Hand Wash
Skincare Soaps and Skin Cleansers
SoftCIDE soap is designed to be extra mild on your hands despite frequent washing
You can wash with SoftCIDE many times a day without it being harsh on your skin and SoftCIDEs unique formulation helps replenish the skins natural oils that are washed away by frequent washing. SoftCIDE is also an antimicrobial. It contains 1% chloroxylenol (PCMX), a fast acting broad spectrum antimicrobial ingredient that is effective against Gram-negative and Gram-positive bacteria as well as yeast.