96 Well Pad, Diversified Biotech
Supplier: Diversified Biotech
WPNP-1000 WPNP-2000 WPNP-3000
490007-138BX 26.32 USD
490007-138 490007-140 490007-142
96 Well Pad, Diversified Biotech
Well Plate Stands and Well Orienters
A note pad that corresponds to the layout of a 96-well plate. A fast and easy way to keep accurate records of samples in a microplate.
The adhesive backing allows the researcher to affix individual sheets into a notebook. Letter and numerical well indicators run along an X and Y axis. Individual cell spaces are left blank, but can be written in to facilitate identification.
Delivery information: The pad contains (50) 5 x 7 sheets.