AluminaSeal
Supplier: DIVERSIFIED BIOTECH
Ideal for cold storage, moderate and high temperature incubation, protection of light sensitive samples and high throughput samples, Strong 2.7 mil thick pierceable aluminum film,Non-cytotoxic and sterile, temperature stable from -80 degree C to 110 degree C.