Wire Marker Card, Brady
Supplier: Brady Worldwide
114052
490006-396PK 118.39 USD
490006-396
Wire Marker Card, Brady
Labels
Repositionable vinyl cloth wire markers from Brady® are perfect for general wire marking needs.
  • Card-mounted wire markers allow for quick, on-the-spot wire marking
  • Excellent flexibility for wrapping around curved surfaces
  • Withstands moderate heat, oil, water, and dirt
Holds tightly to surfaces but allows item to be repositioned with ease if necessary.
Certifications: ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155.