Boekel Serological Pipette Cans
Supplier: Boekel
1366SO
490006-052EA 56.8 USD
490006-052
Boekel Serological Pipette Cans
Pipettes Serological Pipettes
Pipette canisters are fabricated from either stainless steel or a new space-age Ultem® 1000 plastic. Suited for sterilizing pipettes either by autoclaving or with dry heat. Stainless steel boxes offer high resistance to corrosion. Square style eliminates the possibility of pipette cans rolling off the bench. Plastic cans offer transparent visibility of contents, are easy to clean, and virtually indestructible. Plastic pipette cans have a flat edge to prevent rolling.
  • Stainless steel offers high resistance to corrosion
  • Square style eliminates the possibility of pipette cans rolling off the bench
  • Plastic cans offer transparent visibility of contents, are easy to clean, and virtually indestructible
  • Plastic pipette cans have a flat edge to prevent rolling
  • Stainless steel pipette cans feature bonded silicone pads on inside top and bottom to protect against pipette chipping