Magnesium chloride 50 mM in aqueous solution

Supplier: MERIDIAN LIFE SCICNE, INC BE

BIO-37026
490005-950EA 86.42 USD
490005-950
Magnesium chloride 50 mM in aqueous solution
Magnesium chloride

Composition, MgCl2 50 mM in nuclease free water. The 50 mM MgCl2 Solution should be stored at -20°C.

Formula: MgCl₂
Storage Temperature: Ambient
MDL Number: MFCD00011106
CAS Number: 7786-30-3
EINECS: 232-094-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR