BioMix™ Red DNA Polymerase Mix, Bioline Reagents
Supplier: MERIDIAN LIFE SCICNE, INC BE
BIO-25006
490005-934EA 295.71 USD
490005-934
BioMix™ Red DNA Polymerase Mix, Bioline Reagents
Nucleic Acid Reagents End-point PCR Enzymes and Kits
BioMix™ Red is a complete ready-to-use 2x reaction mix containing an ultra-stable Taq DNA polymerase
  • Versatile
  • High throughput and convenient
  • Direct gel loading
  • Suitable for routine PCR applications and TA cloning
It contains an additional inert red dye that permits easy visualization and direct loading onto a gel.
Delivery information: BioMix Red is supplied with additional MgCl2 solution and any fine adjustments be required.