Benchmark BeadBug™ Homogenizer
Supplier: Ward's
D1030
490005-814EA 1579.67 USD
490005-814
Benchmark BeadBug™ Homogenizer
Homogenizers Handheld Homogenizers
The BeadBug is a high energy bench-top homogenizer that sets the performance standard for personal sized cell disruption and lysis instruments.
  • Extremely powerful mixing action, for lysis, grinding, or homogenization
  • Eliminates the cross-contamination associated with hand-held homogenizers
  • Faster and more effective than tissue grinders
  • Mix 1, 2, or 3 tubes simultaneously
  • 2 year warranty
  • Timer 3 seconds to 3 minutes
Simultaneous homogenization of up to 3 samples takes place (often within 45 seconds) inside the disposable 2 mL screw cap microtubes. The optimized mixing motion causes rapid cell disruption through constant high velocity impact from the hardened micro-beads chosen specifically for your sample type.
Developed for samples too tough to be homogenized in a common lab vortexer, the BeadBug provides a convenient and economical alternative to the traditional, over-priced homogenizers on the market today. With up to three tube capacity and a foot print of just 7" x 8", the BeadBug is the ideal homogenizing instrument for low to medium throughput laboratories.
Operating temperature: 4 to 65 °C.