SP Bel-Art Mono-Block™ 12-Well Reservoir, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F37880-1200
490005-652EA 46.29 USD
490005-652
SP Bel-Art Mono-Block™ 12-Well Reservoir, Bel-Art Products, a part of SP
Reagent Reservoirs
Use this reservoir with 8- or 12-channel pipettors for plate washing or feeding.
  • Stable base
  • V-shaped bottom to improve concentration
  • Reduced waste from well design
Each separate well holds 5 mL and has a v-shaped bottom to concentrate fluids and reduce waste. A wide base is stable for bench top use or comfortable to hold in the hand. Mono-Block is a trademark of Bel-Art Scienceware.