Axygen® Tecan Genesis-Style Liquid Level Sensing Filter Pipette Tips, Corning
Supplier: Corning
TTF-1000-CBK-HTR-S
490005-374CS 581.92 USD
490005-374
Axygen® Tecan Genesis-Style Liquid Level Sensing Filter Pipette Tips, Corning
Pipette Tips
Axygen robotic tips are engineered to the exact specifications of the original equipment manufacturers' tips, ensuring complete compatibility and consistent fit with the most popular liquid handling systems.
  • Packaged in plastic blister packs
  • Tips are certified RNase-, DNase-, and pyrogen-free
  • Conductive, sterile
Genesis-style tips are compatible with Tecan Genesis Freedom, Freedom EVO, and MiniPrep robotic workstations, as well as Cavro, PerkinElmer/Packard Multiprobe II HT, and Multiprobe II HT EX workstations.
Tecan, Genesis Freedom, Freedom EVO and Cavro are registered trademarks of Tecan Group, AG. PerkinElmer, Packard and MultiProbe are registered trademarks of PerkinElmer, Inc. Axygen is a registered trademark of Axygen, Inc.