ExpellPlus™ Filter Tips
Supplier: OXFORD LAB PRODUCTS INC
5030030C
490005-252CS 1103.16 USD
490005-252
ExpellPlus™ Filter Tips
Pipette Tips
The Expell tips without the plus low retention properties are of the same high manufacturing standards, maintaining strict tolerances.
  • No Detectable Nucleic acid contamination
  • No Detectable PCR Inhibitors
  • No Detectable Endonucleases
  • No Detectable Endotoxin
  • Sterile
Ideal wetting properties and high transparency and purity levels.