Contact Dish With Grid, Bioplast Manufacturing
Supplier: Akro-Mils
2911
490005-222CS 335.93 USD
490005-222
Contact Dish With Grid, Bioplast Manufacturing
Culture Dishes Petri Dishes
Guaranteed Sterilized via Gamma Irradiation Process. Precision & Convenience. Designed to determine bio-burden counts on working surfaces. Grid consists of 16 (10x10 mm) squares marked numerically in one direction and alphabetically in the other. Media well permits dish and media to be inverted and placed in contact with working surface. Unique outer ring construction prevents contamination from fingers touching the media. Suitable for all automated filling equipment.
  • Stackable construction
  • Made for easy handling
  • Ventilation ribs minimize condensation
  • Domed base reduces media costs
  • Guaranteed sterile via gamma irradiation
  • Disposable