GeneMate Full-Skirted, Low-Profile, 96-Well PCR Plates, Scientific Specialties
Supplier: GeneMate
3441-00-520IS
490003-844BX 127.6 USD
490003-844
GeneMate Full-Skirted, Low-Profile, 96-Well PCR Plates, Scientific Specialties
Microplates PCR Plates
Compatible with most commercially available thermal cyclers.
  • Full-skirt design reduces warping caused by thermal cycling
  • Ultra-thin wall for optimum heat transfer and higher yield
  • High contrast black alphanumerics for rapid sample identification
  • Plates can be sealed with strip caps, sealing mats, or sealing film
  • Crystal-clear clarity
  • Easy to visualize content of each well
  • Certified free from DNase, RNase, and detectable DNA