GeneMate PCR Tubes with Attached Caps, 8-Strip 0.2 ml, Scientific Specialties
Supplier: GeneMate
3230-00-210IS
490003-672BX 349.2 USD
490003-672
GeneMate PCR Tubes with Attached Caps, 8-Strip 0.2 ml, Scientific Specialties
Tubes PCR Tubes
This system virtually ensures the elimination of a major source of contaminationcaps being placed on other than their original tube strips after cycling.
  • Caps need not be replaced due to confusion as to their original orientation on the tubes, accidental dropping, or simply being misplaced
Ordering and inventorying is simplified no more calculating quantities of cap strips to tube strips.
Ordering information: Minimum order requirement of 3 cases.