GeneMate Combination Packs, 12-Strip Standard Tubes with 12-Strip Optically-Clear Flat Caps, Scientific Specialties
Supplier: GeneMate
490003-628
490003-628BX 191.99 USD
490003-628
GeneMate Combination Packs, 12-Strip Standard Tubes with 12-Strip Optically-Clear Flat Caps, Scientific Specialties
Tubes PCR Tubes
Designed to precisely fit most thermal cyclers. Ideal for qPCR (real-time PCR). Convenient tube and cap combination packs. Equal number of tubes and caps in one pack.
  • Secure evaporation-resistant seal between cap lid and tube
  • Optically-clear flat caps
  • Fluorescent signal is passed through unaffected
  • Ultra-thin wall for optimum heat transfer and higher yield
  • Tubes are pre-numbered for easy well identification
  • Crystal-clear clarity, easy to visualize content of each well
  • Available in white
Ideal for enhancing signal strength for qPCR assays.
Certified free DNase, RNase, and detectable DNA.
Ordering information: GeneMate 96-well PCR tube storage racks are available separately.