GeneMate Capless Graduated Microcentrifuge Tubes, 1.7 ml, Scientific Specialties
Supplier: VWR International
490003-284
490003-284BX 115.66 USD
490003-284
GeneMate Capless Graduated Microcentrifuge Tubes, 1.7 ml, Scientific Specialties
Tubes Microcentrifuge Tubes
Our popular microcentrifuge tubes are now available without caps. Best for use in fraction collection and whenever storage of samples is not necessary. Capless tubes are graduated and are available in natural only.
  • Rated 20,000 x g
  • Polypropylene