GeneMate Clearview Co-Polymer 1.7 ml Graduated Microcentrifuge Tubes, Scientific Specialties
Supplier: VWR International
490003-250 490003-261
490003-250BX 138.88 USD
490003-250 490003-261
GeneMate Clearview Co-Polymer 1.7 ml Graduated Microcentrifuge Tubes, Scientific Specialties
Tubes Microcentrifuge Tubes
Made from Co-Polymer, offering high chemical resistance.
  • Easy to open
  • Ultimate in clarity, glass-like
  • Positive seal design (-90˚C to 120˚C)
  • Chamfered tube opening for easy & rapid cap closure
  • Rated to 20,000 x g
  • Limited boiling applications