Aluminum PCR Sealing Foil, GeneMate
Supplier: GeneMate
490001-214
490001-214PK 224.98 USD
490001-214
Aluminum PCR Sealing Foil, GeneMate
Seals Microplate Seals
Ideal for PCR applications, storage of sample material and active agents.
  • Adheres well to polypropylene, polystyrene, and polycarbonate without special equipment
  • Compatible with aqueous solutions and organic solvents
  • Minimal evaporation and cross contamination
  • Temperature resistant from −80 °C to 110 °C