GeneMate Orbital Shaker Mini, Benchmark Scientific
Supplier: Ward's
BT302
490000-126EA 3399.48 USD
490000-126
GeneMate Orbital Shaker Mini, Benchmark Scientific
Shakers and Mixers Orbital Shakers
The Orbital Shaker Mini is a personal sized orbital shaker including a 9.5" x 8" non-slip rubber platform, ideal for tissue culture flasks, petri dishes and staining trays.
  • 2-year full replacement warranty
  • Perfectly circular 19 mm orbit for aeration and mixing
  • Digital speed control up to 300 rpm
  • Equipped with 9.5" x 8" non-skid rubber coated platform
Additional platforms are also available for the Orbital Shaker Mini including the convenient MAGic Clamp™ platform for use with Erlenmeyer flasks and test tube racks. The unique, magnetic attachment method is the easiest way to instantly change between flask clamps of different sizes. Mixing speed and run time are displayed on the large LED control panel located on the front side of the unit. The integral microprocessor includes a constant monitoring system that ensures accuracy through the duration of the program.Sophisticated over-temperature and over-speed controls ensure long life, safety and sample integrity.