GeneMate GyroMixer, Variable Speed, Benchmark Scientific
Supplier: Ward's
GM2300
490000-120EA 2148.6 USD
490000-120
GeneMate GyroMixer, Variable Speed, Benchmark Scientific
Shakers and Mixers Rotating/Rocking Shakers
This mixer is designed to provide precise speeds and tilt angles required for a broad range of molecular and biological mixing applications. The user can adjust both parameters to match the vessel size and the volume of the liquid being mixed, yielding optimum results.
  • 2-year full replacement warranty
  • Nutating motion for uniform mixing
  • Dimpled mat, perfect for centrifuge tubes
  • Incubator and cold room safe
This mixer can accept stackable platforms (clearance= 2.75"). All platform mats are autoclavable. This model has maintenance free brushless motors and are safe for use in cold rooms and incubators.
The rocking speed and tilt angle are both easily adjustable in seconds. At low speeds and minimal tilt angles, this model provides a gentle wave effect for non-foaming mixing, as isrequired for most blot washing and gel work. When set to higher speeds and more extreme tilt angles, it produces aggressive agitation for vigorous sample mixing.