GeneMate Orbital Shaker MP4, Benchmark Scientific
Supplier: Ward's
GM1502
490000-114EA 1606.64 USD
490000-114
GeneMate Orbital Shaker MP4, Benchmark Scientific
Shakers and Mixers Orbital Shakers
The Orbital Shaker MP4 is designed with a 3 mm mixing orbit, optimized for thorough mixing of microplates and/or PCR plates. Up to 4 plates (standard or deep well) can be placed on the included platform and are instantly secured without the use springs, clamps or tools.
  • 2-year full replacement warranty
  • 3 mm orbit, optimized for thorough mixing in microplates
  • Digital speed control up to 1500 rpm
  • Holds up to four microplates
A digital microprocessor ensures accurate speed control over the brushless, maintenance free motor drive and both speed and time can be viewed simultaneously on the large LED display.