J.T.Baker® Robotic Tips Type ST
Supplier: Ritter, part of Avantor
49000-00001 49000-11061 49001-00001 49001-11061 49000-00061 49001-01061
49000-0000CS 2795.66 USD
49000-0000 49000-1106 49001-0000 49001-1106 49000-0006 49001-0106
J.T.Baker® Robotic Tips Type ST
Pipette Tips
Suitable for use on Euroimmun Analyzer. Available in conductive version, with or without filter.
  • Manufactured under cleanroom conditions (cleanroom class 8)
  • Conductive tips for liquid level sensing
  • Ideal for dosing even small volumes with high precision and without contamination
J.T.Baker® robotic tips type ST are suitable for use on Euroimmun Analyzer and other robotic instruments using the same pipetting head.
The validation of the tips on the instrument for specific applications is within the responsibility of the user.