Burundum™ Cylinders, U.S. Stoneware
Supplier: US Stoneware
BRUN125-90 BRUN050-90
48908-068CS 1673.26 USD
48908-068 48908-080
Burundum™ Cylinders, U.S. Stoneware
Mills
For wet or dry processes, the cylindrical shape provides additional contacts for faster, finer grinding
The non-porous surface is hard and chip-resistant, non-contaminating. High specific gravity (3.4) ups “packing density” for most efficient mill use. The smaller size is for liquid dispersion, and the larger size for crushing. Hardness is 9 on the Mohs scale.