Slide Grip, Peel-A-Way® Plastic, Polysciences
Supplier: Polysciences Inc
19801A-1
48440-003PK 127.27 USD
48440-003
Slide Grip, Peel-A-Way® Plastic, Polysciences
Slide Holders
Slide grip is ideal for staining procedures
It is molded from a special plastic compound which tolerates staining chemicals and solvents, even special metal stains. Fits all Coplin and round-mouth staining jars. Holds up to five slides. Withstands 80°C (176°F) temperatures, to eliminate removing slides for drying.