SP Bel-Art Slide Mailer, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H44200-0000
48428-029CS 825.39 USD
48428-029
SP Bel-Art Slide Mailer, Bel-Art Products, a part of SP
Slide Mailers
Chemically inert polyethylene container includes a deep screw cap to provide maximum protection against breakage.
  • Polyethylene construction
  • Chemically inert
  • Easy to grip and clean when projected out of jar
Once the cap is removed, slides project 12.7 mm (½") out of the jar for easy grip and clean handling. Internal grooves accept 5 to 10 microscope 25x75 mm (1x3") slides, or can be used as a specimen jar to hold vials up to 12 mL. The cap has a small loop for attachment of wired tags and should be taped for shipment.