Superfrost® Cytogenetics Slide, Erie Scientific (Thermo Scientific)
Supplier: Erie Scientific
CGN-4951-006
48404-468CS 1130.26 USD
48404-468
Superfrost® Cytogenetics Slide, Erie Scientific (Thermo Scientific)
Slides Microscope Slides
Superfrost® slide feature a glass surface specifically processed and inspected for cytogenetic applications.
Slide features include consistent, high-quality surface for reliable chromosome spreads, hydrophilic surface improves retention of metaphase cells and Superfrost® white tab for permanent marking.