Superforst™ Plus Gold Microscope Slides, Epredia™
Supplier: Erie Scientific
FT4981IGLPLUS-001
48382-200CS 1070.88 USD
48382-200
Superforst™ Plus Gold Microscope Slides, Epredia™
Slides Microscope Slides
Slides feature an adhesive treatment that first attracts and then chemically bonds fresh or formalin-fixed tissue sections securely to the slide surface.
Slides are recommended for hard-to-hold tissue samples such as bone, brain, and breast that are 5 µm or larger. Ideal for special stains, immunocytochemical, and in situ DNA hybridization applications. Slides are frosted with a gold tint.