Polysine™ Adhesion Microscope Slides, Epredia™
Supplier: Erie Scientific
P4981-001
48382-117CS 992.51 USD
48382-117
Polysine™ Adhesion Microscope Slides, Epredia™
Slides Adhesive Slides
Slides provide superior cell and tissue adhesion due to a permanent bio-adhesive material that electrostatically and chemically attracts tissues and tissue sections
These slides are particularly effective in applications involving frozen and paraffin embedded sections, cytocentrifuge preparations, and cytology smears. Not affected by chemicals, enzyme predigestion, or heating. Size: 25x75mm.