Steris Spordex Culture Media
Supplier: STERIS CORP
NA114
48350-012PK 514.4 USD
48350-012
Steris Spordex Culture Media
Media for Microbiology Prepared Media for Microbiology
This media used with Steris Spordex and Spordi biological indicator strips consists of a modified soybean casein digest broth that contains a pH indicator.
  • Consists of a modified soybean casein digest broth that contains a pH indicator
  • Easy-to-read color change in the medium
This media used with Steris Spordex and Spordi biological indicator strips consists of a modified soybean casein digest broth that contains a pH indicator. An easy-to-read color change in the medium to yellow is indicative of microbial growth.