Purospher® STAR HPLC Columns and Cartridges, MilliporeSigma
Supplier: MilliporeSigma
1.50244.0001 1.50245.0001 1.50248.0001 1.50250.0001 1.50251.0001 1.50252.0001 1.50267.0001 1.50358.0001 1.50225.0001 1.50231.0001 1.50240.0001 1.50241.0001 1.50242.0001 1.50249.0001 1.50268.0001 1.50356.0001 1.50357.0001 1.50272.0001 1.50031.0001 1.50032.0001 1.50033.0001 1.50035.0001 1.51453.0001 1.51454.0001 1.50036.0001 1.50037.0001 1.51455.0001 1.50274.0001 1.50038.0001 1.50255.0001 1.50253.0001 1.51460.0001 1.51456.0001 1.50269.0001 1.50253.0001 1.50253.0001 1.50623.0001 1.50624.0001 1.50625.0001 1.50626.0001 1.50627.0001 1.50254.0001 1.50256.0001 1.51918.0001 1.50629.0001 1.50630.0001 1.51013.0001 1.51014.0001 1.51911.0001 1.51912.0001 1.51913.0001 1.51914.0001 1.51916.0001 1.51917.0001 1.50617.1003 1.50648.1003 1.51455.1003 1.51456.1003 1.51918.1003 1.50036.1003 1.50037.1003 1.50251.1003 1.50252.1003 1.50358.1003 1.50359.1003 1.50750.0001
48219-470EA 730.61 USD
48219-470 48219-472 48219-474 48219-476 48219-478 48219-480 48219-482 48219-484 48219-486 48219-488 48219-754 48219-760 48219-768 48219-770 48219-772 48219-782 48219-784 48219-787 48219-789 48219-794 48219-796 48219-798 48219-800 48219-803 48219-805 48219-806 48219-808 48219-810 48219-812 48219-817 48219-821 48219-825 48219-828 48219-870 48219-814 48219-869 71001-970 EM1.50253.0001 EM1.50623.0001 EM1.50624.0001 EM1.50625.0001 EM1.50626.0001 EM1.50627.0001 10143-548 10143-550 10143-994 10143-996 10811-818 10811-820 10811-824 10811-826 10811-838 10811-840 10811-842 10811-844 10811-846 10811-848 10811-850 10811-930 10811-936 10811-950 10811-952 10811-954 10811-956 10811-984 10811-986 10811-998 10812-184 10812-186 10812-188 EM1.50750.0001
Purospher® STAR HPLC Columns and Cartridges, MilliporeSigma
Chromatography Columns
Columns are for use with basic, neutral, metal, chelating, or any other sample format, and are ideal for the separation of difficult hydrophobic samples.
Purospher™ STAR columns are designed to provide optimum results even when loaded with large or small sample volumes and are also available in a narrow-bore format (1 mm). The wide pH range provides the required pH stability for 99% of the common analysis applications. These columns provide the power to separate even the most difficult of compound mixtures. With up to 150000 plates per meter for the 3 µm particle size and 100000 plates per meter for the 5 µm particle size, Purospher™ STAR columns ensure good baseline separations. Special derivatization and endcapping processes ensure long-term chemical stability, excellent retention stability, and long column lifetime. All Purospher™ STAR columns are guaranteed to provide absolutely reproducible results.
Ordering information: All LiChroCART® cartridges require end fittings. manuCART® and LiChroCART® series end fittings and holders are available separately. Hibar® columns include end fittings. If 4×4 mm guard columns are to be used on 4 or 4.6 mm Int.Ø. Hibar® Purospher™ STAR columns, the top fitting must be replaced with guard column holder designed to accommodate this size guard column. Other larger Int.Ø. Purospher™ STAR columns include end fittings as indicated in their descriptions. Guard columns for these larger Int.Ø columns are also available, contact your Avantor sales representative to order.