Trak 300 HEMATRAK Immersion Oil, Thermo Scientific
Supplier: Epredia
M8000
48218-686EA 535.85 USD
48218-686
Trak 300 HEMATRAK Immersion Oil, Thermo Scientific
Stains Microscopy and staining reagents
Specifically designed for HEMATRAK and Micro 21 automated differential systems
  • Safe to use with valuable optics
  • Allows 99% light transmission throughout most of UV-VIS spectrum
  • Contains no PCBs
Oil is standardized at 23 °C to have a refractive index of 1.5150 and viscosity of 300cs.