Digital Salt Meter, Conductivity Method, Model ES-421, ATAGO®
Supplier: Atago
4211
47752-898EA 2503.83 USD
47752-898
Digital Salt Meter, Conductivity Method, Model ES-421, ATAGO®
Electrochemistry Meters Salinity Meters
This digital salt meter has a funnel-shaped sensor section, which measures the salt concentration of a sample by the conductivity method
It is ideal for measuring dilute solution of samples, such as soy sauce, ketchup, mayonnaise, Worcester sauce, and pickle juice.
Ordering information: One-year manufacturer’s warranty. Supplied with a 006P dry battery (9V). A calibration certificate is available. Contact your Avantor representative for information.