Cryogenic Transfer Hose, Airgas
Supplier: Airgas
Y154CH429538
47752-588EA 1066.79 USD
47752-588
Cryogenic Transfer Hose, Airgas
Tubing Transfer Tubing
Hose features stainless steel anti-kink armor casing.
  • Suitable for helium, nitrogen and argon
CGA-end features a 90° bend for ease in connecting to liquid cylinders.