SP Bel-Art Poly-Jaque™ Lab Support Jack, Polypropylene, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F18391-0001
47751-794EA 600.92 USD
47751-794 47751-796
SP Bel-Art Poly-Jaque™ Lab Support Jack, Polypropylene, Bel-Art Products, a part of SP
Laboratory Jacks
Lab support jack supports gradient makers, hot plates, flasks, beakers, baths and other lab items up to 6.8kg (15 lb).
  • Polypropylene construction
  • Smooth and easy-to-clean top plate
  • Not intended to lift weight from bottom position
  • Plates may be drilled for permanent attachent to lab equipment
Molded of strong polycarbonate and assembled with no metal parts, the plastic jack is electrically non-conductive, can be immersed in a bath or used in corrosive environments and is steam autoclavable at 121ºC (250ºF)
The smooth, easy-to-clean top plate has a 21 cm (8.25") diameter and a molded-in position to install a vertical support rod (sold separately). The 46 cm (18") long stainless steel support rod attaches to the top plate to allow attachment of various clamps and other accessories. The rod diameter is 7.9 mm (⁵/₁₆") and is supplied with two hex nuts for secure attachment. Non-corroding, polypropylene burette clamp (sold separately) will not obscure graduations.
Jack is not intended to lift weight from bottom position. Place object on top plate only after jack is cranked to desired height.
A large height adjusting knob allows accurate positioning as an elevated stand from 7.6 cm (3") to 30 cm (12"). Both top and bottom plates may be drilled for permanent attachment to bench or lab equipment with screws or bolts.
Dimensions: Closed 7.6 x 16.5 x 16 cm (3 x 61/2 x 63/16"), lab support jack only"