SP Bel-Art Coplin Staining Jar, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F44208-1000
47751-792EA 10.58 USD
47751-792
SP Bel-Art Coplin Staining Jar, Bel-Art Products, a part of SP
Jars Staining Jars
This polypropylene staining jar improves on the classic tapered square design with a larger opening for easier placement and removal of slides.
  • Polypropylene construction
  • Larger opening than classic tapered square jars
  • Easy to place and remove slides
  • Autoclavable
The polypropylene screw cap is 53 mm in diameter. It has a flat top, easy-grip large knurled edge, deep thread, and a seal ring to prevent leaks. Five internal slots hold up to 10 standard 25x75 mm or 1x3" slides back to back. Jar is steam autoclavable at 121°C (250°F). Remove cap when autoclaving.