SP Bel-Art ProCulture Mini Cryocooler, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F18846-0020
47751-730EA 170.83 USD
47751-730
SP Bel-Art ProCulture Mini Cryocooler, Bel-Art Products, a part of SP
Coolers Benchtop Coolers
This small cooler safely holds twelve 0.5, 1.5 or 2.0 ml microcentrifuge tubes or cryogenic vials at a temperature below –15 ºC for approximately 1 hour. Store at –20 ºC for 24 hours prior to use.
  • Polycarbonate cooler body is filled with a non-toxic gel which cannot contact tubes; ensures markings and labels on tubes are safe
  • Clear polycarbonate lid shows positions of tubes; keyed to correspond with positions printed on cooler
  • Molded side handles allow secure grip for transfer
  • Use included insert to hold 0.5 ml microcentrifuge tubes
  • Rubber feet prevent sliding on the lab bench
  • Stackable
Ideal for protecting valuable cell stocks kept at low temperatures for viability.