Heavyweight Hycar Apron, Ansell
Supplier: Ansell Healthcare
950290
47751-518CS 425.37 USD
47751-518
Heavyweight Hycar Apron, Ansell
Workwear
SType 56-512 aprons are heavyweight for use in chemical processing, food processing, and industrial maintenance.
  • Rugged fabrication
  • Extra-long ties and grommets for secure fit
They are made from Hycar material, a tough nitrile blend, and weigh 510.3g (18oz.). These aprons feature extra-long ties and grommets for closure.