VWR® Spec-Wipe® 4 Presaturated Wipers
Supplier: VWR International
47751-462
47751-462CS 576.09 USD
47751-462
VWR® Spec-Wipe® 4 Presaturated Wipers
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
These Spec-Wipes are excellent for cleaning rough, abrasive, or irregular surfaces, yet soft enough for delicate surfaces.
  • 100% white interlock knit polyester
  • Presaturated
  • Continuous filament
Designed for critical-process cleaning and environments where low particle and fiber generation is critical.
Appropriate for electronics, pharmaceutical, biotechnology, chemical and petroleum, environmental, and education industries. Made from 100% white interlock knit polyester that has been laundered in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom to further reduce particulate and extractable contaminants.
Wipers are presaturated with semiconductor grade solvent to allow for safe storage and dispensing of cleaning solvents while reducing hazardous waste. Durable interlock knit allows for rigorous cleaning. Continuous filament creates softness for delicate surfaces.
Specifications reflect the data for the wiper in an unsaturated state. Wipers are manufactured utilizing IPA (isopropyl alcohol) filtered to 5 µm.
Certifications: Laundered in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom.
Packaging: Half-folded wipers are packaged in a resealable pouch.