SP Bel-Art Burette Clamp, Spring Action, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H18354-0000
47750-930EA 110.02 USD
47750-930
SP Bel-Art Burette Clamp, Spring Action, Bel-Art Products, a part of SP
Clamps General Purpose Clamps
Firmly holds test-tubes, burettes and other labware in place.
  • Spring-action design
  • Stainless steel construction
  • Tightened with a flat wing-head screw
  • Covered in rubber tubing for increased friction
The spring action, stainless steel jaws have a flat wing-head screw for tightening, are covered with rubber tubing for increased friction and a soft grip, and have an opening range of 0 to 25 mm (1").