SP Bel-Art Bosshead, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H18350-0000
47750-928EA 77.97 USD
47750-928
SP Bel-Art Bosshead, Bel-Art Products, a part of SP
Clamps Bossheads
This open-sided stainless steel bosshead accepts rods up to 13 mm (½") diameter.
  • Stainless steel construction
  • Open-sided design
  • Compatible with rods up to 13 mm in diameter
Rods are held at right angles to one another and secured with flat wing-head clamping screws.