SP Bel-Art Leveling Table, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H18310-0000
47750-926EA 288.23 USD
47750-926
SP Bel-Art Leveling Table, Bel-Art Products, a part of SP
Surface Protectors Bench Protectors
This clear and rigid acrylic level surface helps in pouring immunological plates, horizontal agarose gels, and acrylamide sequencing gels.
  • Clear, rigid acrylic construction
  • Adjustable legs
  • Inset leveling bulb assists in accurate adjustment
Each leg is adjustable, and an inset leveling bulb below the working surface assists in accurate adjustment.