SP Bel-Art Poxygrid® Lead Flask Ring Holder, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F18307-1000
47750-924EA 43.16 USD
47750-924
SP Bel-Art Poxygrid® Lead Flask Ring Holder, Bel-Art Products, a part of SP
Support Stands
Wire formed, epoxy-coated holder keeps lead ring weights organized and handy.
  • Epoxy-coated steel wire construction
  • Compact circular base
  • Autoclavable
Occupying very little space, the circular base is only 19 cm (7½") in diameter.
Rings having an inside diameter of 17.8 mm (0.7") or larger stack neatly on a tapered center post which is 17.1 cm (6¾") high. Steam autoclavable at 121˚C (250˚F).