SP Bel-Art Hydrometer Jar, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H17817-0000
47750-918EA 96.26 USD
47750-918
SP Bel-Art Hydrometer Jar, Bel-Art Products, a part of SP
Cylinders Hydrometer Cylinders
Hydrometer Jar is composed of two pieces - a clear, polycarbonate cylinder and a removable hexagonal polypropylene base.
  • Clear polycarbonate cylinder
  • Polypropylene base
  • Broad base prevents tipping
Clarity of polycarbonate cylinder makes reading easy
Hydrometer not included.
Broad polyethylene base helps prevent tipping. The clear, unbreakable polycarbonate cylinder has a 245 mL capacity and the 4” polyethylene base is removable.