SP Bel-Art Heavy-Duty, Long Stem Funnel, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F14671-0000
47750-826PK 37.73 USD
47750-826
SP Bel-Art Heavy-Duty, Long Stem Funnel, Bel-Art Products, a part of SP
Funnels Filtration Funnels Filter Funnels
Heavy-duty, polypropylene plastic funnel with smooth walls, 60-degree cone angle and long stem.
  • Polypropylene construction
  • Smooth walls
  • External ribs
  • Autoclavable
External ribs prevent air lock. Steam autoclavable at 121ºC (250ºF).