SP Bel-Art Fluorescent Ruler, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F13355-0001
47750-776EA 28.93 USD
47750-776
SP Bel-Art Fluorescent Ruler, Bel-Art Products, a part of SP
Rulers
This plastic metric/English scale is printed with a highly visible, glow-in-the-dark pink phosphorescent ink.
  • Highly visible
  • Glow-in-the-dark scale
  • Ideal for viewing fluorescent images and photography
Under UV light the ruler graduations glow visibly and help to calculate nucleic acid mobility. The ruler is marked 0 to 15 cm in millimeter increments, along one edge and 0 to 6" in 1/16" increments on the other edge.