SP Bel-Art Mini Vacuum Desiccator, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F41099-0000
47750-242EA 156.2 USD
47750-242
SP Bel-Art Mini Vacuum Desiccator, Bel-Art Products, a part of SP
Desiccators Desiccators, Plastic
The small size of this desiccator allows quick vacuum creation for rapid, repeated degassing of multiple samples.
  • Compact design
  • Clear polycarbonate top
  • White polypropylene bottom
  • Easy to view contents
Clear polycarbonate top fits securely into the white polypropylene bottom and allows easy viewing of contents. The stopcock with 2 mm Teflon® PTFE plug accepts tubing with ¼" to ⅜” I.D. A peripheral flange around the bottom fits into two snap locks and seals the two halves against a Buna-N O-ring. Desiccator has a vacuum rating of 740 mm Hg (29").