Thick Chromatography Paper, Grade 243, Ahlstrom
Supplier: Ahlstrom
2438-4657
47749-850PK 1109.84 USD
47749-850
Thick Chromatography Paper, Grade 243, Ahlstrom
Blotting Paper
Ahlstrom Grade 243 is one of the thicker chromatography papers available from Ahlstrom
It offers moderately high flow and absorption and is recommended for chromatography with heavy solute loading.