Thin Chromatography Paper, Grade 631, Ahlstrom
Supplier: Ahlstrom
6318-4657
47749-848PK 338.43 USD
47749-848
Thin Chromatography Paper, Grade 631, Ahlstrom
Blotting Paper
Ahlstrom Grade 631 is a think chromatography paper with a faster flow rate
It is recommended for routine chromatography where loading is relatively low.