SP Bel-Art Evaporating Dish, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F40925-0000
47748-186PK 68.78 USD
47748-186
SP Bel-Art Evaporating Dish, Bel-Art Products, a part of SP
Trays/Dishes/Pots Evaporating Basins/Dishes
Smooth polypropylene dish with easy pour spout holds up to 175 mL.
  • Polypropylene construction
  • Reusable
  • Autoclavable
Useful for titration and evaporation, the dish resists acids and alkalis and is autoclavable at 121°C (250°F). It can also be used as a reusable weighing dish.