Standard Incubators with Natural Convection, BD 400
Supplier: Binder
9010-0176
47746-766EA 6731.3 USD
47746-766
Standard Incubators with Natural Convection, BD 400
Incubators Standard Incubators
This incubator is specially designed for gentle incubation of organisms, as well as conditioning of heat-sensitive media or microbiological heating.
  • APT.line™ preheating chamber technology
  • Adjustable exhaust air flap
  • Controller with timer functions
  • Inner door made of tempered safety glass
  • Two chrome-plated racks
  • Class 3.1 independent temperature safety device (DIN 12880) with visual temperature alarm
Safe and reproducible incubation even under high batch throughputs in long-term operation. This give a disinfection routine at 100 °C and provides stable atmosphere by inner glass door.